missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubrene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132380010

 View more versions of this product

Catalog No. AC132380010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Rubrene
517-51-1
Red
98.5% min. (HPLC)
C42H28
1 g
rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
YYMBJDOZVAITBP-UHFFFAOYSA-N
5,6,11,12-tetraphenyltetracene
68203
99%
99%
330°C to 335°C
Authentic
Glass bottle
MFCD00003703
05, II, 726
Solubility in water: practically insoluble
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
532.67
532.67
Crystalline Powder

Chemical Identifiers

517-51-1
532.67
YYMBJDOZVAITBP-UHFFFAOYSA-N
68203
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
C42H28
MFCD00003703
rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
5,6,11,12-tetraphenyltetracene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-242-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only