missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl[(S,S)-Tsdpen](mesitylene) 90.0+%, TCI America™
$87.60 - $288.94
Chemical Identifiers
| CAS | 174813-81-1 |
|---|---|
| Molecular Formula | C30H34ClN2O2RuS |
| Molecular Weight (g/mol) | 623.194 |
| MDL Number | MFCD12545952 |
| InChI Key | BABSEZBJIPZYQB-XCPIVNJJSA-M |
| Synonym | chloro 1s,2s-n-p-toluenesulfonyl-1,2-diphenylethanediamine mesitylene ruthenium ii |
| PubChem CID | 131675785 |
| IUPAC Name | N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0126200MG
|
TCI America
R0126200MG |
200 mg |
Each for $87.60
|
|
|||||
|
R01261G
|
TCI America
R01261G |
1 g |
Each for $288.94
|
|
|||||
Chemical Identifiers
| 174813-81-1 | |
| 623.194 | |
| BABSEZBJIPZYQB-XCPIVNJJSA-M | |
| 131675785 | |
| CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+] |
| C30H34ClN2O2RuS | |
| MFCD12545952 | |
| chloro 1s,2s-n-p-toluenesulfonyl-1,2-diphenylethanediamine mesitylene ruthenium ii | |
| N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride |
Specifications
| 174813-81-1 | |
| MFCD12545952 | |
| chloro 1s,2s-n-p-toluenesulfonyl-1,2-diphenylethanediamine mesitylene ruthenium ii | |
| CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+] | |
| 623.194 | |
| 622.19 | |
| Crystalline Powder |
| C30H34ClN2O2RuS | |
| 200 mg | |
| BABSEZBJIPZYQB-XCPIVNJJSA-M | |
| N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride | |
| 131675785 | |
| ≥90% (HPLC) | |
| RuCl[(S,S)-Tsdpen](mesitylene) |
Safety and Handling
TSCA : No