missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl[(S,S)-Tsdpen](p-cymene) 90.0+%, TCI America™
$102.52 - $329.94
Chemical Identifiers
| CAS | 192139-90-5 |
|---|---|
| Molecular Formula | C31H35ClN2O2RuS |
| MDL Number | MFCD09953484 |
| Synonym | [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0124200MG
|
TCI America
R0124200MG |
200 mg |
Each for $102.52
|
|
|||||
|
R01241G
|
TCI America
R01241G |
1 g |
Each for $329.94
|
|
|||||
Chemical Identifiers
| 192139-90-5 | |
| C31H35ClN2O2RuS | |
| MFCD09953484 | |
| [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) |
Specifications
| 192139-90-5 | |
| MFCD09953484 | |
| [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) | |
| ≥90% (HPLC) | |
| RuCl[(S,S)-Tsdpen](p-cymene) |
| C31H35ClN2O2RuS | |
| 200 mg | |
| 636.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No