missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 195461000

 View more versions of this product

Catalog No. AC195461000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Black
5% Ruthenium
1.02
MFCD00011207 MFCD03458417
01,983; 05,574
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
5% (Ru on dry basis)(typically)
Powder
Ruthenium on activated carbon
7440-18-8,7440-44-0,7732-18-5
Ru
100 g
15, 8433
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
101.07
Glass bottle

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only