Learn More
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution, Thermo Scientific Chemicals
$656.34 - $656.34
Chemical Identifiers
CAS | 34513-98-9 |
---|---|
Molecular Formula | N4O10Ru |
Molecular Weight (g/mol) | 317.09 |
MDL Number | MFCD00016313 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC365041000
|
Thermo Scientific Chemicals
365041000 |
100 mL | Glass bottle |
Each for $656.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34513-98-9 | |
N4O10Ru | |
317.09 | |
MFCD00016313 |
Specifications
Brown to Orange | |
Solution | |
N4O10Ru | |
Solubility in water: soluble | |
317.09 | |
Glass bottle | |
1.0700g/mL |
34513-98-9 | |
100 mL | |
MFCD00016313 | |
317.09 | |
1.5% (Ru) (typically) | |
1.07 | |
Ruthenium(III) nitrosyl nitrate, In diluted nitric acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
May intensify fire; oxidizer.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 252-068-8
RUO – Research Use Only