missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals

$301.32 - $4782.77

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4487903
View Documents
Thermo Scientific Chemicals
04487903
1 g
Each for $301.32
 
AA4487906
View Documents
Thermo Scientific Chemicals
04487906
5 g
Each for $1,143.19
 
AA4487914
View Documents
Thermo Scientific Chemicals
04487914
25 g
Each for $4,782.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]

Specifications

Gray
gray powder
Ru
1 g
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
-200 Mesh
Powder
7440-18-8
>99.99% (Metals basis excluding Ca)
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
Odorless
Ruthenium
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-127-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only