missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
$434.63 - $434.63
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1399404
|
Thermo Scientific Chemicals
01399404 |
2 g |
Each for $434.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Specifications
Gray | |
gray powder | |
Ru | |
2 g | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
-325 Mesh | |
Ruthenium |
7440-18-8 | |
99.9% (Metals basis) | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only