missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Rutin trihydrate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1357036

 View more versions of this product

Catalog No. AAA1357036


Explore available promotions

Description

Description

Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

Solubility
Soluble in DMSO, acetone, dichloromethane or ethyl acetate. Insoluble in water.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

(+)-Rutin trihydrate
184°C (decomposition)
MFCD00149490
75455
14,3802
Soluble in DMSO,acetone,dichloromethane or ethyl acetate. Insoluble in water.
+9° (c=0.5 in Ethanol)
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
16218542
97%
250249-75-3
C27H36O19
500 g
Light sensitive
rutoside trihydrate, +-rutin trihydrate, rutin trihydrate, rutin hydrate, unii-rf4n03853g, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-3-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyltetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy-4h-chromen-4-one trihydrate, rutoside trihydrate ep, rutin hydrate jan, rutin trihydrate hplc, rutin trihydrate, analytical standard
NLLBWFFSGHKUSY-JPRRWYCFSA-N
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)O)C5=CC(=C(C=C5)O)O)O)O)O)O)O)O.O.O.O
664.566
664.58 (610.52 Anhydrous)

Chemical Identifiers

250249-75-3
664.566
NLLBWFFSGHKUSY-JPRRWYCFSA-N
16218542
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)O)C5=CC(=C(C=C5)O)O)O)O)O)O)O)O.O.O.O
C27H36O19
MFCD00149490
rutoside trihydrate, +-rutin trihydrate, rutin trihydrate, rutin hydrate, unii-rf4n03853g, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-3-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyltetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy-4h-chromen-4-one trihydrate, rutoside trihydrate ep, rutin hydrate jan, rutin trihydrate hplc, rutin trihydrate, analytical standard
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-814-1

RTECSNumber : VM2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only