missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1'-Bi-2-naphthol 98.0+%, TCI America™

$180.81 - $544.30

Chemical Identifiers

CAS 18531-99-2
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00004068
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11005G
View Documents
TCI America
B11005G
5 g
Each for $180.81
Only null left
 
B110025G
View Documents
TCI America
B110025G
25 g
Each for $544.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

18531-99-2
286.33
PPTXVXKCQZKFBN-UHFFFAOYSA-N
11762
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
C20H14O2
MFCD00004068
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol

Specifications

18531-99-2
White-Yellow
MFCD00004068
2811
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
11762
≥98.0% (HPLC,T)
(S)-(-)-1,1′-Bi-2-naphthol
210°C
C20H14O2
5 g
1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
286.33
286.33
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1807

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'