missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L20242MD

 View more versions of this product

Catalog No. AAL20242MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate
1.118
93°C (199°F)
1.6045
250 mg
4675069
s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate
+47° (c=3.5 in Toluene)
1-[(1S)-1-isocyanatoethyl]naphthalene
7023510
95%
73671-79-1
106°C to 108°C (0.16 mmHg)
C13H11NO
MFCD00064181
UN2206
Moisture sensitive; May form a precipitate
GONOHGQPZFXJOJ-UHFFFAOYNA-N
CC(N=C=O)C1=C2C=CC=CC2=CC=C1
197.24
197.24

Chemical Identifiers

73671-79-1
197.24
GONOHGQPZFXJOJ-UHFFFAOYNA-N
7023510
CC(N=C=O)C1=C2C=CC=CC2=CC=C1
C13H11NO
MFCD00064181
s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate
1-[(1S)-1-isocyanatoethyl]naphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only