missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98.0+%, TCI America™
$111.84 - $328.08
Chemical Identifiers
| CAS | 225920-05-8 |
|---|---|
| Molecular Formula | C10H8F6O |
| Molecular Weight (g/mol) | 258.163 |
| MDL Number | MFCD03093010 |
| InChI Key | MMSCIQKQJVBPIR-YFKPBYRVSA-N |
| Synonym | (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol |
| PubChem CID | 9816575 |
| IUPAC Name | (1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol |
| SMILES | CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)O |
Chemical Identifiers
| 225920-05-8 | |
| 258.163 | |
| MMSCIQKQJVBPIR-YFKPBYRVSA-N | |
| 9816575 | |
| CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)O |
| C10H8F6O | |
| MFCD03093010 | |
| (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol | |
| (1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol |
Specifications
| 225920-05-8 | |
| White | |
| MFCD03093010 | |
| (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol | |
| CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)O | |
| 258.163 | |
| 258.16 | |
| Crystalline Powder |
| 54°C | |
| C10H8F6O | |
| 1 g | |
| MMSCIQKQJVBPIR-YFKPBYRVSA-N | |
| (1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol | |
| 9816575 | |
| ≥98.0% (GC) | |
| (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol |
Safety and Handling
TSCA : No