missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5243506

 View more versions of this product

Catalog No. AAH5243506


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(S)-1-Boc-3-(hydroxymethyl)piperidine
C11H21NO3
5 g
Slightly soluble in water.
CC(C)(C)OC(=O)N1CCCC(C1)CO
215.293
215.29
140695-84-7
MFCD02683203
s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine
OJCLHERKFHHUTB-VIFPVBQESA-N
tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate
1514448
97%

Chemical Identifiers

140695-84-7
215.293
OJCLHERKFHHUTB-VIFPVBQESA-N
1514448
CC(C)(C)OC(=O)N1CCCC(C1)CO
C11H21NO3
MFCD02683203
s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine
tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions