missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2707814

 View more versions of this product

Catalog No. AAH2707814


Explore available promotions

Description

Description

(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases. Light sensitive.
Specifications

Specifications

(S)-(+)-1-Bromo-2-methylbutane
534-00-9
121°C to 122°C
C5H11Br
MFCD00078662
UN1993
Light sensitive; May form a precipitate
s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s
XKVLZBNEPALHIO-YFKPBYRVSA-N
CC[C@H](C)CBr
151.05
151.05
stabilized with potassium carbonate
1.223
22°C (79°F)
1.4455
25 g
1718815
14,601
Insoluble in water.
+4.5° (Neat)
(2S)-1-bromo-2-methylbutane
5464167
97%

Chemical Identifiers

534-00-9
151.05
XKVLZBNEPALHIO-YFKPBYRVSA-N
5464167
CC[C@H](C)CBr
C5H11Br
MFCD00078662
s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s
(2S)-1-bromo-2-methylbutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 208-583-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only