missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1-Methoxy-2-propanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 331790050

 View more versions of this product

Catalog No. AC331790050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-(+)-1-Methoxy-2-propanol
98.5
-97.0°C
0.9200g/mL
30°C
98.5% min. (GC)
C4H10O2
CH3OCH2CH(OH)CH3
5 mL
+20° (20°C c=10,CHCl3)
s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
ARXJGSRGQADJSQ-BYPYZUCNSA-N
(2S)-1-methoxypropan-2-ol
6993459
99%
26550-55-0
100.0
Colorless
120.0°C
Authentic
Glass bottle
1.4020 to 1.4040
MFCD01632588
0.92
+ 20.00
Solubility in water: soluble
CC(COC)O
90.12
90.12
Liquid

Chemical Identifiers

26550-55-0
90.12
ARXJGSRGQADJSQ-BYPYZUCNSA-N
6993459
CC(COC)O
C4H10O2
MFCD01632588
s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
(2S)-1-methoxypropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only