missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol 95.0+%, TCI America™

$439.92 - $2632.05

Chemical Identifiers

CAS 161511-85-9
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD03093620
InChI Key XIRUXUKRGUFEKC-UHFFFAOYNA-N
Synonym (S)-1-Boc-2-azetidinemethanol
PubChem CID 10511797
IUPAC Name tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B2123100MG
View Documents
TCI America
B2123100MG
100 mg
Each for $439.92
Only null left
 
B21231G
View Documents
TCI America
B21231G
1 g
Each for $2,632.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

161511-85-9
187.24
XIRUXUKRGUFEKC-UHFFFAOYNA-N
10511797
CC(C)(C)OC(=O)N1CCC1CO
C9H17NO3
MFCD03093620
(S)-1-Boc-2-azetidinemethanol
tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate

Specifications

161511-85-9
C9H17NO3
(S)-1-Boc-2-azetidinemethanol
CC(C)(C)OC(=O)N1CCC1CO
100 mg
10511797
≥95.0% (GC)
(S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol
Yellow
MFCD03093620
XIRUXUKRGUFEKC-UHFFFAOYNA-N
tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate
187.24
187.24
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'