missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™

$61.22 - $90.86

Chemical Identifiers

CAS 101469-92-5
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD01317839
InChI Key APCBTRDHCDOPNY-ZETCQYMHSA-N
Synonym s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
PubChem CID 854055
IUPAC Name tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30551G
View Documents
TCI America
B30551G
1 g
Each for $61.22
 
B30555G
View Documents
TCI America
B30555G
5 g
Each for $90.86
 
Description

Description

Specifications

Chemical Identifiers

101469-92-5
187.239
APCBTRDHCDOPNY-ZETCQYMHSA-N
854055
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD01317839
s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate

Specifications

101469-92-5
White-Yellow
MFCD01317839
s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
CC(C)(C)OC(=O)N1CCC(C1)O
187.239
187.24
Crystalline Powder
63°C
C9H17NO3
1 g
APCBTRDHCDOPNY-ZETCQYMHSA-N
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
854055
≥98.0% (GC)
(S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'