missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

$989.79 - $989.79

Chemical Identifiers

CAS 1365531-94-7
Molecular Formula C52H60O8P2
Molecular Weight (g/mol) 874.992
MDL Number MFCD19443626
InChI Key PKHREKHNGLJKHV-UHFFFAOYSA-N
Synonym 2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
PubChem CID 87110691
IUPAC Name [2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6083203
View Documents
Thermo Scientific Chemicals
H6083203
1 g
Each for $989.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1365531-94-7
874.992
PKHREKHNGLJKHV-UHFFFAOYSA-N
87110691
CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
C52H60O8P2
MFCD19443626
2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
[2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane

Specifications

1365531-94-7
MFCD19443626
Air Sensitive
PKHREKHNGLJKHV-UHFFFAOYSA-N
[2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane
87110691
≥97%
C52H60O8P2
1 g
2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC
874.992
874.98
(S)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only