Learn More
(S)-(+)-2-Butanol, 98+%
CAS: 4221-99-2 | C4H10O | 74.123 g/mol
$124.62 - $467.81
Chemical Identifiers
| Molecular Formula | C4H10O |
|---|---|
| Molecular Weight (g/mol) | 74.123 |
| MDL Number | MFCD00064281 |
| InChI Key | BTANRVKWQNVYAZ-BYPYZUCNSA-N |
| Synonym | s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol |
| PubChem CID | 444683 |
| ChEBI | CHEBI:45475 |
| IUPAC Name | (2S)-butan-2-ol |
| SMILES | CCC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1416403
|
Thermo Scientific Chemicals
L1416403 |
1 g |
Each for $124.62
|
|
|||||
|
AAL1416406
|
Thermo Scientific Chemicals
L1416406 |
5 g |
Each for $467.81
|
|
|||||
Description
For synthesis of optically active products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H10O | |
| MFCD00064281 | |
| s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol | |
| CHEBI:45475 | |
| CCC(C)O |
| 74.123 | |
| BTANRVKWQNVYAZ-BYPYZUCNSA-N | |
| 444683 | |
| (2S)-butan-2-ol |
Specifications
| 4221-99-2 | |
| 99°C to 100°C | |
| 26°C (79°F) | |
| 1.398 | |
| MFCD00064281 | |
| 1718763 | |
| s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol | |
| CCC(C)O | |
| 74.123 | |
| CHEBI:45475 | |
| ≥98% |
| 0.806 | |
| 1 g | |
| Alcohol-like | |
| C4H10O | |
| UN1120 | |
| 141541 | |
| BTANRVKWQNVYAZ-BYPYZUCNSA-N | |
| (2S)-butan-2-ol | |
| 444683 | |
| 74.12 | |
| (S)-(+)-2-Butanol |
Safety and Handling
GHS H Statement
H226-H319-H335-H336
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber : 224-168-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only