missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2-Chloropropionic acid, 98%, Thermo Scientific Chemicals

$119.78 - $330.53

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0942706
View Documents
Thermo Scientific Chemicals
L0942706
5 g
Each for $119.78
 
AAL0942714
View Documents
Thermo Scientific Chemicals
L0942714
25 g
Each for $330.53
 
Description

Description

Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

Solubility
Fully miscible in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

29617-66-1
107.51
GAWAYYRQGQZKCR-REOHCLBHSA-M
107915
(2S)-2-chloropropanoic acid
C3H4ClO2
MFCD00064205
s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
CHEBI:73956
C[C@H](Cl)C([O-])=O

Specifications

29617-66-1
1.27
107°C (225°F)
1.4345
5 g
1720257
Fully miscible in water.
C[C@H](Cl)C([O-])=O
107.51
CHEBI:73956
99%
4°C
187°C
C3H4ClO2
MFCD00064205
UN2511
s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
GAWAYYRQGQZKCR-REOHCLBHSA-M
(2S)-2-chloropropanoic acid
107915
108.53
(S)-(-)-2-Chloropropionic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: 2-CHLOROPROPIONIC ACID

EINECSNumber : 411-150-5

RTECSNumber : UA2451950

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only