Learn More
(S)-(-)-2-Methylbutylamine, 98+%, Thermo Scientific Chemicals
$265.79 - $265.79
Chemical Identifiers
CAS | 34985-37-0 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD00064430 |
InChI Key | VJROPLWGFCORRM-YFKPBYRVSA-N |
Synonym | s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride |
PubChem CID | 2724272 |
IUPAC Name | (2S)-2-methylbutan-1-amine |
SMILES | CCC(C)CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1406303
|
Thermo Scientific Chemicals
L1406303 |
1 g |
Each for $265.79
|
|
|||||
Description
(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
Chemical Identifiers
34985-37-0 | |
87.166 | |
VJROPLWGFCORRM-YFKPBYRVSA-N | |
2724272 | |
CCC(C)CN |
C5H13N | |
MFCD00064430 | |
s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride | |
(2S)-2-methylbutan-1-amine |
Specifications
34985-37-0 | |
96°C to 97°C | |
C5H13N | |
MFCD00064430 | |
UN2733 | |
Air Sensitive | |
Fully miscible in water. | |
−5.9° (Neat) | |
(2S)-2-methylbutan-1-amine | |
2724272 | |
≥98% |
0.756 | |
3°C (37°F) | |
1.4105 | |
1 g | |
1718807 | |
s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride | |
VJROPLWGFCORRM-YFKPBYRVSA-N | |
CCC(C)CN | |
87.166 | |
87.16 | |
(S)-(-)-2-Methylbutylamine |
Safety and Handling
GHS H Statement
H225-H314-H318-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 252-307-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only