missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-2-Phenylpropionic acid, 97%, Thermo Scientific Chemicals
$129.67 - $359.55
Chemical Identifiers
CAS | 7782-24-3 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.177 |
MDL Number | MFCD00063139 |
InChI Key | YPGCWEMNNLXISK-ZETCQYMHSA-N |
Synonym | s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc |
PubChem CID | 2724622 |
ChEBI | CHEBI:48527 |
IUPAC Name | (2S)-2-phenylpropanoic acid |
SMILES | CC(C1=CC=CC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL08802MD
|
Thermo Scientific Chemicals
L08802MD |
250 mg |
Each for $129.67
|
|
|||||
AAL0880203
|
Thermo Scientific Chemicals
L0880203 |
1 g |
Each for $359.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-24-3 | |
150.177 | |
YPGCWEMNNLXISK-ZETCQYMHSA-N | |
2724622 | |
(2S)-2-phenylpropanoic acid |
C9H10O2 | |
MFCD00063139 | |
s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc | |
CHEBI:48527 | |
CC(C1=CC=CC=C1)C(=O)O |
Specifications
7782-24-3 | |
1.1 | |
148°C (298°F) | |
1.523 | |
250 mg | |
s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc | |
+73° (c=1.6 in Chloroform) | |
(2S)-2-phenylpropanoic acid | |
2724622 | |
150.18 | |
(S)-(+)-2-Phenylpropionic acid |
30°C to 34°C | |
264°C to 266°C | |
C9H10O2 | |
MFCD00063139 | |
2044507 | |
YPGCWEMNNLXISK-ZETCQYMHSA-N | |
CC(C1=CC=CC=C1)C(=O)O | |
150.177 | |
CHEBI:48527 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only