Learn More
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1171122
Description
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.
Solubility
Soluble in water (10-15% at 20°C), alcohol, acetone, dimethyl sufoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid | |
158°C to 162°C | |
MFCD00005272 | |
82132 | |
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
OC(=O)C1CCC(=O)N1 | |
129.12 | |
CHEBI:18183 | |
≥98% |
98-79-3 | |
C5H7NO3 | |
100 g | |
14,8001 | |
Soluble in water (10-15% at 20°C),alcohol,acetone,dimethyl sufoxide and methanol. | |
−10.5° (c=5 in Water) | |
(2S)-5-oxopyrrolidine-2-carboxylic acid | |
7405 | |
129.12 |
Chemical Identifiers
98-79-3 | |
129.12 | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
7405 | |
OC(=O)C1CCC(=O)N1 |
C5H7NO3 | |
MFCD00005272 | |
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo | |
CHEBI:18183 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
EINECSNumber : 202-700-3
RTECSNumber : TW3710000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only