missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals

$54.21 - $120.75

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
PubChem CID 7405
ChEBI CHEBI:18183
IUPAC Name (2S)-5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132120250
SDS View Documents
Thermo Scientific Chemicals
132120250
25 g Plastic bottle
Each for $54.21
 
AC132121000
SDS View Documents
Thermo Scientific Chemicals
132121000
100 g Plastic bottle
Each for $120.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-79-3
129.12
ODHCTXKNWHHXJC-UHFFFAOYNA-N
7405
(2S)-5-oxopyrrolidine-2-carboxylic acid
C5H7NO3
MFCD00005272
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
CHEBI:18183
OC(=O)C1CCC(=O)N1

Specifications

98-79-3
White
98%
C5H7NO3
25 g
15,8113
− 27.50
Solubility in water: 100-150g/l (20°). Other solubilities: soluble in alcohol and acetone
OC(=O)C1CCC(=O)N1
129.12
CHEBI:18183
98%
(S)-(-)-2-Pyrrolidone-5-carboxylic acid
152.0°C to 162.0°C
Authentic
Plastic bottle
MFCD00005272
22,284
− 27.50 (20.00°C c=10,1 N NaOH)
l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
ODHCTXKNWHHXJC-UHFFFAOYNA-N
(2S)-5-oxopyrrolidine-2-carboxylic acid
7405
129.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 202-700-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only