missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-3-[1-(Dimethylamino)ethyl]phenol 98.0+%, TCI America™
$89.35 - $176.98
Chemical Identifiers
CAS | 139306-10-8 |
---|---|
Molecular Formula | C10H15NO |
Molecular Weight (g/mol) | 165.24 |
MDL Number | MFCD06656491 |
InChI Key | GQZXRLWUYONVCP-UHFFFAOYNA-N |
PubChem CID | 445892 |
IUPAC Name | 3-[1-(dimethylamino)ethyl]phenol |
SMILES | CC(N(C)C)C1=CC(O)=CC=C1 |
Chemical Identifiers
139306-10-8 | |
165.24 | |
GQZXRLWUYONVCP-UHFFFAOYNA-N | |
3-[1-(dimethylamino)ethyl]phenol |
C10H15NO | |
MFCD06656491 | |
445892 | |
CC(N(C)C)C1=CC(O)=CC=C1 |
Specifications
139306-10-8 | |
White | |
MFCD06656491 | |
GQZXRLWUYONVCP-UHFFFAOYNA-N | |
3-[1-(dimethylamino)ethyl]phenol | |
445892 | |
≥98.0% (GC,T) | |
(S)-3-[1-(Dimethylamino)ethyl]phenol |
119°C | |
C10H15NO | |
1 g | |
CC(N(C)C)C1=CC(O)=CC=C1 | |
165.24 | |
165.24 | |
Crystalline Powder |
Safety and Handling
TSCA : No