missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-3-(Benzoylthio)-2-methylpropionic Acid 98.0+%, TCI America™
$84.78 - $84.78
Chemical Identifiers
CAS | 72679-02-8 |
---|---|
Molecular Formula | C11H12O3S |
Molecular Weight (g/mol) | 224.274 |
MDL Number | MFCD00038376 |
InChI Key | BCAYPPFBOJCRPN-MRVPVSSYSA-N |
PubChem CID | 12763916 |
IUPAC Name | (2S)-3-benzoylsulfanyl-2-methylpropanoic acid |
SMILES | CC(CSC(=O)C1=CC=CC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B53151G
|
TCI America
B53151G |
1 g |
Each for $84.78
|
|
|||||
Chemical Identifiers
72679-02-8 | |
224.274 | |
BCAYPPFBOJCRPN-MRVPVSSYSA-N | |
(2S)-3-benzoylsulfanyl-2-methylpropanoic acid |
C11H12O3S | |
MFCD00038376 | |
12763916 | |
CC(CSC(=O)C1=CC=CC=C1)C(=O)O |
Specifications
72679-02-8 | |
White-Yellow | |
MFCD00038376 | |
BCAYPPFBOJCRPN-MRVPVSSYSA-N | |
(2S)-3-benzoylsulfanyl-2-methylpropanoic acid | |
12763916 | |
≥98.0% (GC,T) | |
(S)-3-(Benzoylthio)-2-methylpropionic Acid |
69°C | |
C11H12O3S | |
1 g | |
CC(CSC(=O)C1=CC=CC=C1)C(=O)O | |
224.274 | |
224.27 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator