missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-3-Benzyloxy-1,2-propanediol 98.0+%, TCI America™
$59.65 - $350.44
Chemical Identifiers
| CAS | 17325-85-8 |
|---|---|
| Molecular Formula | C10H14O3 |
| Molecular Weight (g/mol) | 182.219 |
| MDL Number | MFCD00077914 |
| InChI Key | LWCIBYRXSHRIAP-JTQLQIEISA-N |
| Synonym | (S)-(-)-Glycerol alpha-Benzyl Ether |
| PubChem CID | 6950841 |
| IUPAC Name | (2S)-3-phenylmethoxypropane-1,2-diol |
| SMILES | C1=CC=C(C=C1)COCC(CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2142100MG
|
TCI America
B2142100MG |
100 mg |
Each for $59.65
|
|
|||||
|
B21421G
|
TCI America
B21421G |
1 g |
Each for $350.44
|
|
|||||
Chemical Identifiers
| 17325-85-8 | |
| 182.219 | |
| LWCIBYRXSHRIAP-JTQLQIEISA-N | |
| 6950841 | |
| C1=CC=C(C=C1)COCC(CO)O |
| C10H14O3 | |
| MFCD00077914 | |
| (S)-(-)-Glycerol alpha-Benzyl Ether | |
| (2S)-3-phenylmethoxypropane-1,2-diol |
Specifications
| 17325-85-8 | |
| C10H14O3 | |
| 100 mg | |
| LWCIBYRXSHRIAP-JTQLQIEISA-N | |
| (2S)-3-phenylmethoxypropane-1,2-diol | |
| 6950841 | |
| ≥98.0% (GC) | |
| (S)-(-)-3-Benzyloxy-1,2-propanediol |
| White-Yellow | |
| MFCD00077914 | |
| (S)-(-)-Glycerol alpha-Benzyl Ether | |
| C1=CC=C(C=C1)COCC(CO)O | |
| 182.219 | |
| 182.22 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator