Learn More
(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals
$720.93 - $720.93
Chemical Identifiers
CAS | 218943-30-7 |
---|---|
Molecular Formula | C18H25NO6 |
Molecular Weight (g/mol) | 351.40 |
MDL Number | MFCD01862936 |
InChI Key | JABNOCWCLLYHKE-UHFFFAOYNA-N |
Synonym | boc-beta-hoglu obzl-oh, boc-l-beta-homoglutamic acid 6-benzyl ester, boc-beta-homoglu obzl-oh, s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, 3s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, boc-l-beta-homoglutamic acid obzl, ambotzbaa6170, boc-?-hoglu obzl-oh, fmoc-l-beta-homo-glu obzl-oh |
PubChem CID | 2761517 |
IUPAC Name | (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-6-oxo-6-phenylmethoxyhexanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5218703
|
Thermo Scientific Chemicals
H5218703 |
1 g |
Each for $720.93
|
|
|||||
Description
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsStarting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
218943-30-7 | |
351.40 | |
JABNOCWCLLYHKE-UHFFFAOYNA-N | |
2761517 | |
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)CC(O)=O |
C18H25NO6 | |
MFCD01862936 | |
boc-beta-hoglu obzl-oh, boc-l-beta-homoglutamic acid 6-benzyl ester, boc-beta-homoglu obzl-oh, s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, 3s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, boc-l-beta-homoglutamic acid obzl, ambotzbaa6170, boc-?-hoglu obzl-oh, fmoc-l-beta-homo-glu obzl-oh | |
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-6-oxo-6-phenylmethoxyhexanoic acid |
Specifications
218943-30-7 | |
MFCD01862936 | |
boc-beta-hoglu obzl-oh, boc-l-beta-homoglutamic acid 6-benzyl ester, boc-beta-homoglu obzl-oh, s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, 3s-6-benzyloxy-3-tert-butoxycarbonyl amino-6-oxohexanoic acid, boc-l-beta-homoglutamic acid obzl, ambotzbaa6170, boc-?-hoglu obzl-oh, fmoc-l-beta-homo-glu obzl-oh | |
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)CC(O)=O | |
351.40 | |
351.4 | |
1 g |
C18H25NO6 | |
8079168 | |
JABNOCWCLLYHKE-UHFFFAOYNA-N | |
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-6-oxo-6-phenylmethoxyhexanoic acid | |
2761517 | |
95% | |
(S)-3-(Boc-amino)adipic acid 6-benzyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only