Learn More
(S)-(-)-3-Butyn-2-ol, 95%, 98% ee, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 362410010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-(-)-3-Butyn-2-ol | |
2914-69-4 | |
100.0 | |
108°C to 111°C | |
97.5% min. (GC) | |
94% min. (GC) | |
C4H6O | |
1g | |
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc | |
CC(C#C)O | |
70.09 | |
70.09 |
95%, (98% e.e.) | |
94.0 | |
0.8940g/mL | |
26°C | |
Authentic | |
Glass bottle | |
MFCD00190166 | |
0.894 | |
GKPOMITUDGXOSB-BYPYZUCNSA-N | |
(2S)-but-3-yn-2-ol | |
6995470 | |
95%,98% ee |
Chemical Identifiers
2914-69-4 | |
70.09 | |
GKPOMITUDGXOSB-BYPYZUCNSA-N | |
6995470 | |
CC(C#C)O |
C4H6O | |
MFCD00190166 | |
s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc | |
(2S)-but-3-yn-2-ol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Fatal in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Fatal if swallowed.
Flammable liquid and vapor.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
TSCA : TSCA
RUO â Research Use Only