missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 412120050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-3-Hydroxy-3-phenylpropanoic acid | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00145262 | |
−22.50° (20°C c=4,CH3OH) | |
s-3-hydroxy-3-phenylpropanoic acid, 3s-3-hydroxy-3-phenylpropanoic acid, 3s-3-hydroxy-3-phenylpropanoic acid, s-3-hydroxy-3-phenylpropionic acid, ?-3-phenylhydracrylic acid, benzenepropanoic acid,, a-hydroxy-,, as, s---3-hydroxy-3-phenylpropionic acid, 98+%, s---3-hydroxy-3-phenylpropionic acid, 98+%, unii-s2w4lws09c component | |
C1=CC=C(C=C1)C(CC(=O)O)O | |
166.18 | |
CHEBI:51058 | |
99% |
36567-72-3 | |
100.0 | |
99% | |
C9H10O3 | |
5g | |
− 22.50 | |
AYOLELPCNDVZKZ-QMMMGPOBSA-N | |
(3S)-3-hydroxy-3-phenylpropanoic acid | |
2735057 | |
166.18 |
RUO â Research Use Only