missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Hydroxytetrahydrofuran 98.0+%, TCI America™
$29.83 - $70.84
Chemical Identifiers
| CAS | 86087-23-2 |
|---|---|
| Molecular Formula | C4H8O2 |
| Molecular Weight (g/mol) | 88.106 |
| MDL Number | MFCD00064327 |
| InChI Key | XDPCNPCKDGQBAN-BYPYZUCNSA-N |
| Synonym | s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s |
| PubChem CID | 2733227 |
| IUPAC Name | (3S)-oxolan-3-ol |
| SMILES | C1COCC1O |
Chemical Identifiers
| 86087-23-2 | |
| 88.106 | |
| XDPCNPCKDGQBAN-BYPYZUCNSA-N | |
| 2733227 | |
| C1COCC1O |
| C4H8O2 | |
| MFCD00064327 | |
| s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s | |
| (3S)-oxolan-3-ol |
Specifications
| 86087-23-2 | |
| 80°C | |
| Clear Liquid at 20°C | |
| MFCD00064327 | |
| XDPCNPCKDGQBAN-BYPYZUCNSA-N | |
| (3S)-oxolan-3-ol | |
| 2733227 | |
| ≥98.0% (GC) |
| Yellow | |
| 1 g | |
| C4H8O2 | |
| s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s | |
| C1COCC1O | |
| 88.106 | |
| 88.11 | |
| (S)-3-Hydroxytetrahydrofuran |
Safety and Handling
TSCA : No