missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-4-Phenyl-2-oxazolidinone, 99%, Thermo Scientific™

Chemical Identifiers

CAS 99395-88-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00043396
InChI Key QDMNNMIOWVJVLY-MRVPVSSYSA-N
Synonym s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
PubChem CID 730424
IUPAC Name (4S)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC303850010
SDS View Documents
Thermo Scientific Chemicals
303850010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC303850050
SDS View Documents
Thermo Scientific Chemicals
303850050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

99395-88-7
163.18
QDMNNMIOWVJVLY-MRVPVSSYSA-N
730424
C1C(NC(=O)O1)C2=CC=CC=C2
C9H9NO2
MFCD00043396
s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
(4S)-4-phenyl-1,3-oxazolidin-2-one

Specifications

99395-88-7
100.0
98.5% min. (GC)
C9H9NO2
1g
+ 49.5
QDMNNMIOWVJVLY-MRVPVSSYSA-N
(4S)-4-phenyl-1,3-oxazolidin-2-one
730424
99%
98.5
Authentic
Glass bottle
MFCD00043396
+49.5° (20°C c=2,CHCl3)
s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
C1C(NC(=O)O1)C2=CC=CC=C2
163.18
163.18
(S)-(+)-4-Phenyl-2-oxazolidinone, 99%
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only