Learn More
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%
CAS: 17342-08-4 | C5H9NO2 | 115.132 g/mol
$115.99 - $370.71
Identifiants chimiques
| CAS | 17342-08-4 |
|---|---|
| Molecular Formula | C5H9NO2 |
| Molecular Weight (g/mol) | 115.132 |
| MDL Number | MFCD00077792 |
| InChI Key | HOBJEFOCIRXQKH-BYPYZUCNSA-N |
| Synonym | l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847 |
| PubChem CID | 643511 |
| IUPAC Name | (5S)-5-(hydroxymethyl)pyrrolidin-2-one |
| SMILES | C1CC(=O)NC1CO |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAL1835903
|
Thermo Scientific Chemicals
L1835903 |
1 g |
chaque for $115.99
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAL1835906
|
Thermo Scientific Chemicals
L1835906 |
5 g |
chaque for $370.71
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Identifiants chimiques
| 17342-08-4 | |
| 115.132 | |
| HOBJEFOCIRXQKH-BYPYZUCNSA-N | |
| 643511 | |
| C1CC(=O)NC1CO |
| C5H9NO2 | |
| MFCD00077792 | |
| l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847 | |
| (5S)-5-(hydroxymethyl)pyrrolidin-2-one |
Spécifications
| 17342-08-4 | |
| 147°C to 149°C (0.06 mmHg) | |
| MFCD00077792 | |
| 4657914 | |
| Soluble in water. | |
| +30° (c=2 in Ethanol) | |
| (5S)-5-(hydroxymethyl)pyrrolidin-2-one | |
| 643511 | |
| 98% |
| 78°C to 82°C | |
| C5H9NO2 | |
| 1 g | |
| l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847 | |
| HOBJEFOCIRXQKH-BYPYZUCNSA-N | |
| C1CC(=O)NC1CO | |
| 115.132 | |
| 115.13 | |
| (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only