missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-Abscisic Acid 98.0+%, TCI America™
Supplier: TCI America A1698100MG
Specifications
(S)-(+)-Abscisic Acid | |
163°C | |
C15H20O4 | |
100 mg | |
C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O | |
264.32 | |
CHEBI:2365 | |
≥98.0% (HPLC) |
21293-29-8 | |
White-Yellow | |
MFCD00066545,MFCD00066545,MFCD00075619 | |
JLIDBLDQVAYHNE-YKALOCIXSA-N | |
(2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid | |
5280896 | |
264.32 | |
Crystalline Powder |
Chemical Identifiers
21293-29-8 | |
264.32 | |
JLIDBLDQVAYHNE-YKALOCIXSA-N | |
CHEBI:2365 | |
C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O |
C15H20O4 | |
MFCD00066545,MFCD00066545,MFCD00075619 | |
5280896 | |
(2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid |
Safety and Handling
RTECSNumber : RZ2475100
TSCA : No
Recommended Storage : Refrigerator