missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-Benzylsuccinic Acid 98.0+%, TCI America™
$235.03 - $235.03
Chemical Identifiers
CAS | 3972-36-9 |
---|---|
Molecular Formula | C11H12O4 |
Molecular Weight (g/mol) | 208.213 |
MDL Number | MFCD00798341 |
InChI Key | GTOFKXZQQDSVFH-VIFPVBQESA-N |
PubChem CID | 446168 |
IUPAC Name | (2S)-2-benzylbutanedioic acid |
SMILES | C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
B41425G
|
TCI America
B41425G |
5 g |
Each for $235.03
|
|
||||||
Chemical Identifiers
3972-36-9 | |
208.213 | |
GTOFKXZQQDSVFH-VIFPVBQESA-N | |
(2S)-2-benzylbutanedioic acid |
C11H12O4 | |
MFCD00798341 | |
446168 | |
C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O |
Specifications
3972-36-9 | |
White | |
MFCD00798341 | |
GTOFKXZQQDSVFH-VIFPVBQESA-N | |
(2S)-2-benzylbutanedioic acid | |
446168 | |
≥98.0% (HPLC,T) | |
(S)-Benzylsuccinic Acid |
166°C | |
C11H12O4 | |
5 g | |
C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O | |
208.213 | |
208.21 | |
Crystalline Powder |
Safety and Handling
TSCA : No