missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1262214

 View more versions of this product

Catalog No. AAA1262214

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

S-Butyrylthiocholine iodide
172°C to 176°C
CH3CH2CH2COSCH2CH2N(CH3)3I
25 g
Hygroscopic; Light sensitive
WEQAAFZDJROSBF-UHFFFAOYSA-M
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
74630
98%
1866-16-6
C9H20INOS
MFCD00011845
3729509
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
CCCC(=O)SCC[N+](C)(C)C.[I-]
317.229
317.23

Chemical Identifiers

1866-16-6
317.229
WEQAAFZDJROSBF-UHFFFAOYSA-M
74630
CCCC(=O)SCC[N+](C)(C)C.[I-]
C9H20INOS
MFCD00011845
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-475-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only