missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Carboxymethyl-L-cysteine, 97%, Thermo Scientific Chemicals

$82.00 - $220.97

Chemical Identifiers

CAS 638-23-3
Molecular Formula C5H9NO4S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00002614
InChI Key GBFLZEXEOZUWRN-VKHMYHEASA-N
Synonym s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
PubChem CID 193653
ChEBI CHEBI:16163
SMILES N[C@@H](CSCC(O)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2348714
View Documents
Thermo Scientific Chemicals
B2348714
25 g
Each for $82.00
 
AAB2348722
View Documents
Thermo Scientific Chemicals
B2348722
100 g
Each for $220.97
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

638-23-3
179.19
GBFLZEXEOZUWRN-VKHMYHEASA-N
193653
N[C@@H](CSCC(O)=O)C(O)=O
C5H9NO4S
MFCD00002614
s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
CHEBI:16163

Specifications

∼200°C (decomposition)
C5H9NO4S
1725012
s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
N[C@@H](CSCC(O)=O)C(O)=O
25 g
193653
−29° (c=10 in 10% NaHCO)
97%
638-23-3
MFCD00002614
14,1802
GBFLZEXEOZUWRN-VKHMYHEASA-N
(2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
179.19
CHEBI:16163
179.2
S-Carboxymethyl-L-cysteine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-327-5

RTECSNumber : AY4342000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only