missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Curve™ Beta Storage Containers
Ideal for storing reagents that emit beta isotopes
Supplier: S-Curve™ BC502
Description
- 3/8 in. thick (0.95cm) clear acrylic provides a protective beta barrier
- Full-length hinge for durability and strength
- Rubber pads on bottom to prevent sliding and scratching
- Choice of two sizes
Specifications
Rubber pads, flame treated edges | |
For storing reagents that emit beta isotopes | |
Clear | |
30.48cm | |
Acrylic |
12 in. | |
25.4cm | |
Beta storage container | |
10 in. | |
0.37 in. |