S-Curve™ Single Compartment Packaged Glove Dispenser
Specially designed for sterile or packaged gloves, with a single compartment that dispenses gloves in a first-in first-out fashion.
Supplier:  S-Curve™ SD300PETG
 View more versions of this product
Catalog No. 19-170-653
Explore available promotions
Description
Description
  • Specially designed for sterile or pair-packaged gloves
  • Gloves dispense in a first-in, first-out fashion
  • Single, sloped lid with a full-length hinge to maintain HEPA flow
  • Can be wall-mounted with heavy-duty mounting bracket (included)
  • Flame-treated edges for a smooth finish
Specifications
Specifications
Packaged Glove Dispenser
12 x 7 x 17 in.
PETG
Top Loading
Clear
Sterile Pair-Packaged Gloves
1
0.25 in.