missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Ethyl Thiopropionate 98.0+%, TCI America™
$86.90 - $258.73
Chemical Identifiers
CAS | 2432-42-0 |
---|---|
Molecular Formula | C5H10OS |
Molecular Weight (g/mol) | 118.194 |
MDL Number | MFCD00027016 |
InChI Key | HNEVHBHRLCAKKQ-UHFFFAOYSA-N |
Synonym | s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester |
PubChem CID | 75513 |
IUPAC Name | S-ethyl propanethioate |
SMILES | CCC(=O)SCC |
Chemical Identifiers
2432-42-0 | |
118.194 | |
HNEVHBHRLCAKKQ-UHFFFAOYSA-N | |
75513 | |
CCC(=O)SCC |
C5H10OS | |
MFCD00027016 | |
s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester | |
S-ethyl propanethioate |
Specifications
2432-42-0 | |
Colorless | |
C5H10OS | |
5 mL | |
s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester | |
CCC(=O)SCC | |
118.194 | |
118.19 | |
Liquid |
-95°C | |
126°C | |
MFCD00027016 | |
1993 | |
HNEVHBHRLCAKKQ-UHFFFAOYSA-N | |
S-ethyl propanethioate | |
75513 | |
≥98.0% (GC) | |
S-Ethyl Thiopropionate |
Safety and Handling
TSCA : No