Learn More
(S)-(-)-Glycidol, 97%, (98% ee), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 411000250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(S)-(-)-Glycidol | |
60456-23-7 | |
100.0 | |
Colorless to Yellow | |
160°C to 161°C | |
97.5% min. (GC) | |
96% min. (GC) | |
C3H6O2 | |
MFCD00074874 | |
1.116 | |
s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s | |
CTKINSOISVBQLD-VKHMYHEASA-N | |
[(2S)-oxiran-2-yl]methanol | |
6973630 | |
74.08 | |
Liquid |
98% ee | |
96.0 | |
-54°C | |
1.1160g/mL | |
66°C | |
Authentic | |
Glass bottle | |
1.4310 to 1.4340 (20°C, 589nm) | |
25 g | |
−12° to −18° (20°C, 589nm) (neat) | |
Solubility in water: soluble | |
C1C(O1)CO | |
74.08 | |
CHEBI:38690 | |
97%,(98% ee) |
Chemical Identifiers
60456-23-7 | |
74.08 | |
CTKINSOISVBQLD-VKHMYHEASA-N | |
6973630 | |
[(2S)-oxiran-2-yl]methanol |
C3H6O2 | |
MFCD00074874 | |
s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s | |
CHEBI:38690 | |
C1C(O1)CO |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
May damage fertility.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only