missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-Glycidyl 3-Nitrobenzenesulfonate 98.0+%, TCI America™
$143.60 - $422.14
Chemical Identifiers
CAS | 115314-14-2 |
---|---|
Molecular Formula | C9H9NO6S |
Molecular Weight (g/mol) | 259.23 |
MDL Number | MFCD00064582 |
InChI Key | AIHIHVZYAAMDPM-QMMMGPOBSA-N |
Synonym | s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate |
PubChem CID | 146490 |
IUPAC Name | [(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate |
SMILES | [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1 |
Chemical Identifiers
115314-14-2 | |
259.23 | |
AIHIHVZYAAMDPM-QMMMGPOBSA-N | |
146490 | |
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1 |
C9H9NO6S | |
MFCD00064582 | |
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate | |
[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate |
Specifications
115314-14-2 | |
Yellow | |
MFCD00064582 | |
1325 | |
AIHIHVZYAAMDPM-QMMMGPOBSA-N | |
[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate | |
146490 | |
≥98.0% (GC) | |
(S)-Glycidyl 3-Nitrobenzenesulfonate |
65°C | |
C9H9NO6S | |
5 g | |
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate | |
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1 | |
259.23 | |
259.23 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : DB7192010
TSCA : No