missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Glycidyl Phenyl Ether 98.0+%, TCI America™
$68.97 - $210.63
Chemical Identifiers
| CAS | 71031-03-3 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD06659026 |
| InChI Key | FQYUMYWMJTYZTK-SECBINFHSA-N |
| Synonym | (S)-2-(Phenoxymethyl)oxirane, (S)-1,2-Epoxy-3-phenoxypropane |
| PubChem CID | 1534345 |
| IUPAC Name | (2S)-2-(phenoxymethyl)oxirane |
| SMILES | C1C(O1)COC2=CC=CC=C2 |
Chemical Identifiers
| 71031-03-3 | |
| 150.177 | |
| FQYUMYWMJTYZTK-SECBINFHSA-N | |
| 1534345 | |
| C1C(O1)COC2=CC=CC=C2 |
| C9H10O2 | |
| MFCD06659026 | |
| (S)-2-(Phenoxymethyl)oxirane, (S)-1,2-Epoxy-3-phenoxypropane | |
| (2S)-2-(phenoxymethyl)oxirane |
Specifications
| 71031-03-3 | |
| C9H10O2 | |
| 1 g | |
| FQYUMYWMJTYZTK-SECBINFHSA-N | |
| (2S)-2-(phenoxymethyl)oxirane | |
| 1534345 | |
| ≥98.0% (GC) | |
| (S)-Glycidyl Phenyl Ether |
| Colorless | |
| MFCD06659026 | |
| (S)-2-(Phenoxymethyl)oxirane, (S)-1,2-Epoxy-3-phenoxypropane | |
| C1C(O1)COC2=CC=CC=C2 | |
| 150.177 | |
| 150.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0559&(3)-0594
TSCA : No