missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-Mandelic acid, 99+%, Thermo Scientific Chemicals
$42.52 - $430.19
Chemical Identifiers
CAS | 17199-29-0 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00004495 |
InChI Key | IWYDHOAUDWTVEP-ZETCQYMHSA-N |
Synonym | s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid |
PubChem CID | 439616 |
ChEBI | CHEBI:32800 |
IUPAC Name | (2S)-2-hydroxy-2-phenylacetic acid |
SMILES | C1=CC=C(C=C1)C(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1506106
|
Thermo Scientific Chemicals
A1506106 |
5 g |
Each for $42.52
|
|
|||||
AAA1506114
|
Thermo Scientific Chemicals
A1506114 |
25 g |
Each for $133.79
|
|
|||||
AAA1506122
|
Thermo Scientific Chemicals
A1506122 |
100 g |
Each for $430.19
|
|
|||||
Description
(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.
Solubility
Soluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
17199-29-0 | |
152.149 | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
439616 | |
(2S)-2-hydroxy-2-phenylacetic acid |
C8H8O3 | |
MFCD00004495 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
CHEBI:32800 | |
C1=CC=C(C=C1)C(C(=O)O)O |
Specifications
131°C to 135°C | |
C8H8O3 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
(2S)-2-hydroxy-2-phenylacetic acid | |
439616 | |
152.15 | |
5 g | |
Light sensitive |
17199-29-0 | |
MFCD00004495 | |
Soluble in water. | |
C1=CC=C(C=C1)C(C(=O)O)O | |
152.149 | |
CHEBI:32800 | |
≥99% | |
+155° (c=5 in Water) | |
(S)-(+)-Mandelic acid |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
EINECSNumber : 241-240-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only