missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Piperazine-2-carboxylic acid dihydrochloride, 98+%

CAS: 158663-69-5 | C5H12Cl2N2O2 | 203.06 g/mol

$100.45 - $284.22

Chemical Identifiers

CAS 158663-69-5
Molecular Formula C5H12Cl2N2O2
Molecular Weight (g/mol) 203.06
MDL Number MFCD01632104,MFCD03840829
InChI Key WNSDZBQLMGKPQS-UHFFFAOYNA-N
Synonym s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
PubChem CID 2734260
IUPAC Name (2S)-piperazine-2-carboxylic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)C1CNCCN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2897203
View Documents
Thermo Scientific Chemicals
H2897203
1 g
Each for $100.45
Only null left
 
AAH2897206
View Documents
Thermo Scientific Chemicals
H2897206
5 g
Each for $284.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

158663-69-5
203.06
WNSDZBQLMGKPQS-UHFFFAOYNA-N
2734260
Cl.Cl.OC(=O)C1CNCCN1
C5H12Cl2N2O2
MFCD01632104,MFCD03840829
s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
(2S)-piperazine-2-carboxylic acid;dihydrochloride

Specifications

158663-69-5
C5H12Cl2N2O2
1 g
s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
−5° (c=1 in Water)
(2S)-piperazine-2-carboxylic acid;dihydrochloride
2734260
≥98%
∽280°C (decomposition)
MFCD01632104,MFCD03840829
Hygroscopic
WNSDZBQLMGKPQS-UHFFFAOYNA-N
Cl.Cl.OC(=O)C1CNCCN1
203.06
203.07
(S)-(-)-Piperazine-2-carboxylic acid dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only