missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S,S)-DIPAMP, 97%, Thermo Scientific Chemicals
$156.59 - $847.86
Chemical Identifiers
CAS | 97858-62-3 |
---|---|
Molecular Formula | C28H28O2P2 |
Molecular Weight (g/mol) | 458.48 |
MDL Number | MFCD05863547 |
InChI Key | QKZWXPLBVCKXNQ-UHFFFAOYNA-N |
Synonym | 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane |
PubChem CID | 11419748 |
IUPAC Name | (S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane |
SMILES | COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH28037MC
|
Thermo Scientific Chemicals
H28037MC |
100 mg |
Each for $156.59
|
|
|||||
AAH28037MD
|
Thermo Scientific Chemicals
H28037MD |
250 mg |
Each for $311.05
|
|
|||||
AAH2803703
|
Thermo Scientific Chemicals
H2803703 |
1 g |
Each for $847.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97858-62-3 | |
458.48 | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
11419748 | |
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 |
C28H28O2P2 | |
MFCD05863547 | |
1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane | |
(S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane |
Specifications
97858-62-3 | |
C28H28O2P2 | |
100 mg | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1 | |
458.48 | |
458.47 | |
(S,S)-DIPAMP |
101°C to 105°C | |
MFCD05863547 | |
1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane | |
+84° (c=1 in Chloroform) | |
(S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane | |
11419748 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only