missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-Hydrobenzoin 99.0+%, TCI America™

$357.90 - $1,131.48

Chemical Identifiers

CAS 2325-10-2
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064255
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
PubChem CID 853020
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08245G
View Documents
TCI America
H08245G
5 g N/A
Only null left
 
H082425G
View Documents
TCI America
H082425G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2325-10-2
214.26
IHPDTPWNFBQHEB-KBPBESRZSA-N
853020
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
C14H14O2
MFCD00064255
s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
(1S,2S)-1,2-diphenylethane-1,2-diol

Specifications

2325-10-2
White
MFCD00064255
s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
214.26
214.26
Crystalline Powder
148°C
C14H14O2
5 g
IHPDTPWNFBQHEB-KBPBESRZSA-N
(1S,2S)-1,2-diphenylethane-1,2-diol
853020
≥99.0% (GC)
(S,S)-(-)-Hydrobenzoin
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'