3M™ S-Series Hood with Inner Shroud, Integrated Head Suspension
Economy hood for use with 3M™ PAPR and SAR systems. Available in small/medium or medium/large size.
Supplier: 3M™ 7000052786
Description
- Versaflo S-Series economy hood
- Polypropylene
- Air Regulating Valve, Cooling / Heating Valve, Low Pressure Valve
- 20/Case
- Applications include cleaning, assembly, mechanical, painting
- 3M Catalog# S-403L-20, S-403S-20
Specifications
1000 | |
White | |
Hood with Inner Shroud | |
Military maintenance, repair and operation, pharmaceutical, transportation, metal production and fabrication, automotive, food and beverage manufacturing | |
Head, face, neck, and shoulder coverage, fabric suspension, inner shroud | |
20/Cs. | |
PETG |
NIOSH | |
Polypropylene | |
Large | |
Powered air purifying and supplied air respirator systems | |
S-403L-20 | |
Integrated |