Learn More
Thermo Scientific™ Sabouraud Dextrose Agar
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.
$70.74 - $799.23
Specifications
Product Type Agar
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Format Manufacturing Origin Price Quantity  
Catalog Number Mfr. No. Quantity Format Manufacturing Origin Price Quantity  
R01768
View Documents
Thermo Scientific™
R01768
10/Pk. Monoplate Deep Pour United States
Pack of 10 Each for $70.74
Only null left
R112551
View Documents
Thermo Scientific™
R112551
10/Pk. BB - 500mL United States
Case of 10 Each for $799.23
Only null left
R112562
View Documents
Thermo Scientific™
RE112562
10/Pk. BRB - 100mL United States
Pack of 10 Each for $538.14
Only null left
Description
Description
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.
Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
Agar
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'