missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Saccharin Sodium, Dihydrate, Crystalline, USP, 99-101%, Spectrum™ Chemical
SO195, 6155-57-3, C7H4NNaO3S·2H2O
Supplier: Spectrum Chemical Mfg Cor SO19525KG
Description
Spectrum™ Chemical Saccharin Sodium, Dihydrate, Crystalline, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
6155-57-3 | |
Poly Bottle | |
2.5 kg | |
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 | |
241.19 | |
USP |
1 | |
C7H8NNaO5S | |
NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate | |
99 to 101% |
Chemical Identifiers
6155-57-3 | |
241.19 | |
sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate |
C7H8NNaO5S | |
NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |