missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Saccharin, sodium salt hydrate, 99+%, Thermo Scientific Chemicals
$119.01 - $267.23
Chemical Identifiers
CAS | 82385-42-0 |
---|---|
Molecular Formula | C7H4NNaO3S |
Molecular Weight (g/mol) | 205.16 |
MDL Number | MFCD00149605 |
InChI Key | OAZGZMKGRPRHJX-UHFFFAOYSA-M |
Synonym | saccharin sodium salt hydrate |
PubChem CID | 131673955 |
IUPAC Name | 1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate |
SMILES | [Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC223372500
|
Thermo Scientific Chemicals
223372500 |
250 g | Plastic bottle |
Each for $119.01
|
|
||||
AC223370010
|
Thermo Scientific Chemicals
223370010 |
1 kg | Plastic bottle |
Each for $267.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
82385-42-0 | |
205.16 | |
OAZGZMKGRPRHJX-UHFFFAOYSA-M | |
131673955 | |
[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |
C7H4NNaO3S | |
MFCD00149605 | |
saccharin sodium salt hydrate | |
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate |
Specifications
82385-42-0 | |
Authentic | |
Plastic bottle | |
MFCD00149605 | |
27,17 | |
saccharin sodium salt hydrate | |
OAZGZMKGRPRHJX-UHFFFAOYSA-M | |
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate | |
131673955 | |
99+% | |
Saccharin, sodium salt hydrate, 99+% |
Colorless to White | |
25ppm Toluenesulfonamides (GC) max | |
C7H4NNaO3S | |
250 g | |
10,8171 | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol, practically insoluble in diethyl ether and, chloroform | |
[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 | |
205.16 | |
205.16 | |
Crystalline Powder or Crystals |
RUO – Research Use Only